Morpholine-4-carbohydrazide ≥95%
Supplier: Matrix Scientific
085708-1G
102845-780EA
584.62
USD
102845-780
Morpholine-4-carbohydrazide ≥95%
Morpholine-4-carbohydrazide
Hazard: Irritant
Formula:
C₅H₁₁N₃O₂ |
MDL Number:
MFCD01570142 CAS Number: 29053-23-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...