(Benzo[3,4-d]1,3-dioxolan-5-ylmethylene)methane-1,1-dicarbonitrile ≥95%

Supplier: Matrix Scientific

085704-1G 085704-10G
102845-768EA 496.92 USD
102845-768 102845-770
(Benzo[3,4-d]1,3-dioxolan-5-ylmethylene)methane-1,1-dicarbonitrile ≥95%
(Benzo[3,4-d]1,3-dioxolan-5-ylmethylene)methane-1,1-dicarbonitrile

Hazard: Irritant

Formula: C₁₁H₆N₂O₂
MDL Number: MFCD00022987
CAS Number: 2972-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR