4-[2-(Dimethylamino)ethyl]piperazine-1-carboximidamide hydroiodide ≥97%

Supplier: Matrix Scientific

085699-1G
102845-754EA 750.77 USD
102845-754
4-[2-(Dimethylamino)ethyl]piperazine-1-carboximidamide hydroiodide ≥97%
4-[2-(Dimethylamino)ethyl]piperazine-1-carboximidamide hydroiodide

Hazard: Irritant

Formula: C₉H₂₂IN₅
MDL Number: MFCD07368559
CAS Number: 849777-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR