4-Acetyltetrahydro-1(2H)-pyrazinecarboximidamidehydroiodide ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Acetylpiperazine-1-carboxamidine hydroiodide

085698-1G
102845-752EA 298.46 USD
102845-752
4-Acetyltetrahydro-1(2H)-pyrazinecarboximidamidehydroiodide ≥97%
4-Acetyltetrahydro-1(2H)-pyrazinecarboximidamidehydroiodide

Hazard: Irritant

Formula: C₇H₁₅IN₄O
MW: 298.13 g/mol
MDL Number: MFCD06797494
CAS Number: 849776-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR