1-(1-Ethylpropyl)piperazine ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(Pent-3-yl)piperazine

085695-1G 085695-10G
102845-738EA 184.62 USD
102845-738 102845-740
1-(1-Ethylpropyl)piperazine ≥97%
1-(1-Ethylpropyl)piperazine

Hazard: Irritant

Formula: C₉H₂₀N₂
MW: 156.27 g/mol
MDL Number: MFCD02093554
CAS Number: 373356-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR