1-(4-Methylpyridin-2-yl)piperazine ≥97%
Supplier: Matrix Scientific
085683-1G
102845-692EA
327.69
USD
102845-692
1-(4-Methylpyridin-2-yl)piperazine ≥97%
1-(4-Methylpyridin-2-yl)piperazine
Hazard: Irritant
Formula:
C₁₀H₁₅N₃ |
MDL Number:
MFCD03412118 CAS Number: 34803-67-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...