N,N-Diethyl-N-(3-piperazin-1-ylpropyl)amine ≥97%
Supplier: Matrix Scientific
Synonyms:
1-(3-Diethylaminopropyl)piperazine
085679-10G
085679-1G
102845-678EA
646.15
USD
102845-678
102845-676
N,N-Diethyl-N-(3-piperazin-1-ylpropyl)amine ≥97%
N,N-Diethyl-N-(3-piperazin-1-ylpropyl)amine
Hazard: Irritant
Formula:
C₁₁H₂₅N₃ MW: 199.34 g/mol |
MDL Number:
MFCD01075182 CAS Number: 22764-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...