4-Methylpiperazine-1-carboximidamide hydroiodide ≥97%
Supplier: Matrix Scientific
085674-1G
102845-660EA
241.54
USD
102845-660
4-Methylpiperazine-1-carboximidamide hydroiodide ≥97%
4-Methylpiperazine-1-carboximidamide hydroiodide
Hazard: Irritant
Formula:
C₆H₁₅IN₄ |
MDL Number:
MFCD09025920 CAS Number: 77723-03-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...