1-(2-Pyridyl)piperazine monohydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(1-Piperazinyl)pyridine Monohydrochloride

085673-1G
102845-658EA 156.92 USD
102845-658
1-(2-Pyridyl)piperazine monohydrochloride ≥97%
1-(2-Pyridyl)piperazine monohydrochloride

Hazard: Irritant

Formula: C₉H₁₃N₃
MW: 199.68 g/mol
MDL Number: MFCD01861295
CAS Number: 129394-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR