Piperidinepentanenitrile ≥97%

Supplier: Matrix Scientific

085656-1G
102845-614EA 383.08 USD
102845-614
Piperidinepentanenitrile ≥97%
Piperidinepentanenitrile

Hazard: Irritant

Formula: C₁₀H₁₈N₂
MDL Number: MFCD00044836
CAS Number: 4734-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR