1-(Methylsulfonyl)piperidin-4-ol ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Hydroxy-1-(methylsulphonyl)piperidine
085655-1G
102845-612EA
1032.31
USD
102845-612
1-(Methylsulfonyl)piperidin-4-ol ≥97%
1-(Methylsulfonyl)piperidin-4-ol
Hazard: Irritant
Formula:
C₆H₁₃NO₃S MW: 179.24 g/mol |
MDL Number:
MFCD11044218 CAS Number: 141482-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...