cis-2,6-Dimethylpiperidine ≥97%
Supplier: Matrix Scientific
Synonyms:
cis-2,6-Lupetidine
085654-1G
085654-10G
102845-608EA
54.05
USD
102845-608
102845-610
cis-2,6-Dimethylpiperidine ≥97%
cis-2,6-Dimethylpiperidine
Hazard: Irritant
Formula:
C₇H₁₅N |
MDL Number:
MFCD00066304 CAS Number: 766-17-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...