1,4'-Bipiperidine-4'-carboxamide ≥97%

Supplier: Matrix Scientific

085646-1G
102845-584EA 284.62 USD
102845-584
1,4'-Bipiperidine-4'-carboxamide ≥97%
1,4'-Bipiperidine-4'-carboxamide

Hazard: Irritant

Formula: C₁₁H₂₁N₃O
MW: 211.31 g/mol
MDL Number: MFCD01631148
CAS Number: 39633-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR