1-(2-Chloroethyl)piperidine ≥97%
Supplier: Matrix Scientific
085644-1G
085644-10G
102845-578EA
2000
USD
102845-578
102845-576
1-(2-Chloroethyl)piperidine ≥97%
1-(2-Chloroethyl)piperidine
Hazard: Toxic/Corrosive
Formula:
C₇H₁₄ClN Boiling Pt: 69 °C (13 Torr) Melting Pt: 229…230 °C Density: 1.009 |
MDL Number:
MFCD00129177 CAS Number: 1932-03-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...