4-Acetyl-4-phenylpiperidine hydrochloride ≥97%

Supplier: Matrix Scientific

085642-1G 085642-10G
102845-570EA 184.62 USD
102845-570 102845-572
4-Acetyl-4-phenylpiperidine hydrochloride ≥97%
4-Acetyl-4-phenylpiperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₇NO
MDL Number: MFCD00039037
CAS Number: 10315-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR