4-Oxopiperidine-1-carboxamide ≥97%

Supplier: Matrix Scientific

085637-1G
102845-558EA 750.77 USD
102845-558
4-Oxopiperidine-1-carboxamide ≥97%
4-Oxopiperidine-1-carboxamide

Hazard: Irritant

Formula: C₆H₁₀N₂O₂
MDL Number: MFCD00665157
CAS Number: 306976-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR