1-Ethylpiperidin-3-one hydrochloride ≥97%
Supplier: Matrix Scientific
085635-10G
085635-1G
102845-554EA
1163.08
USD
102845-554
102845-552
1-Ethylpiperidin-3-one hydrochloride ≥97%
1-Ethylpiperidin-3-one hydrochloride
Hazard: Irritant
Formula:
C₇H₁₄ClNO |
MDL Number:
MFCD00012793 CAS Number: 41361-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...