tert-Butyl-3-{[3-(trifluoromethyl)phenyl]amino}piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

085632-1G
102845-544EA 835.38 USD
102845-544
tert-Butyl-3-{[3-(trifluoromethyl)phenyl]amino}piperidine-1-carboxylate ≥95%
tert-Butyl-3-{[3-(trifluoromethyl)phenyl]amino}piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₃F₃N₂O₂
MW: 344.38 g/mol
MDL Number: MFCD07786019
CAS Number: 887585-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR