Methyl (4S)-2-oxo-3-piperidin-4-yl-1,3-oxazolidine-4-carboxylate hydrochloride ≥97%

Supplier: Matrix Scientific

085619-1G
102845-516EA 2249.23 USD
102845-516
Methyl (4S)-2-oxo-3-piperidin-4-yl-1,3-oxazolidine-4-carboxylate hydrochloride ≥97%
Methyl (4S)-2-oxo-3-piperidin-4-yl-1,3-oxazolidine-4-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₇ClN₂O₄
MDL Number: MFCD09991839
CAS Number: 1217447-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR