Piperidine-1-carboximidamide hydrochloride ≥97%

Supplier: Matrix Scientific

085612-1G
102845-502EA 298.46 USD
102845-502
Piperidine-1-carboximidamide hydrochloride ≥97%
Piperidine-1-carboximidamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₄ClN₃
MDL Number: MFCD09800604
CAS Number: 41316-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR