1-(4-Fluorobenzoyl)piperidine ≥97%

Supplier: Matrix Scientific

085611-1G 085611-10G
102845-500EA 387.69 USD
102845-500 102845-498
1-(4-Fluorobenzoyl)piperidine ≥97%
1-(4-Fluorobenzoyl)piperidine

Hazard: Irritant

Formula: C₁₂H₁₄BrNO
MDL Number: MFCD00174322
CAS Number: 58547-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR