tert-Butyl-1,4'-bipiperidine-1'-carboxylate ≥97%

Supplier: Matrix Scientific

085600-1G
102845-470EA 1315.38 USD
102845-470
tert-Butyl-1,4'-bipiperidine-1'-carboxylate ≥97%
tert-Butyl-1,4'-bipiperidine-1'-carboxylate

Hazard: Irritant

Formula: C₁₅H₂₈N₂O₂
MW: 268.4 g/mol
MDL Number: MFCD07779459
CAS Number: 125541-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR