4-(3-Pyridyl)-1H-1,2,3-triazole-5-carbonitrile ≥95%

Supplier: Matrix Scientific

085591-1G
102845-450EA 553.85 USD
102845-450
4-(3-Pyridyl)-1H-1,2,3-triazole-5-carbonitrile ≥95%
4-(3-Pyridyl)-1H-1,2,3-triazole-5-carbonitrile

Hazard: Irritant

Formula: C₈H₅N₅
MW: 171.16 g/mol
MDL Number: MFCD00169915
CAS Number: 884504-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR