4-Methyl-2H-1,4-benzoxazin-3(4H)-one ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Methyl-2H-1,4-Benzoxazin-3-one

085582-10G 085582-1G
102845-424EA 287.88 USD
102845-424 102845-422
4-Methyl-2H-1,4-benzoxazin-3(4H)-one ≥97%
4-Methyl-2H-1,4-benzoxazin-3(4H)-one

Hazard: Irritant

Formula: C₉H₉NO₂
MW: 163.18 g/mol
MDL Number: MFCD00601365
CAS Number: 21744-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR