4-[(3-Oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid ≥97%

Supplier: Matrix Scientific

085581-1G
102845-420EA 636.92 USD
102845-420
4-[(3-Oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid ≥97%
4-[(3-Oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid

Hazard: Irritant

Formula: C₁₆H₁₃NO₄
MW: 283.29 g/mol
MDL Number: MFCD00269588
CAS Number: 857492-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR