3,4-Dihydro-2H-1,4-benzoxazine-2-carbothioamide ≥97%

Supplier: Matrix Scientific

085578-1G
102845-414EA 426.15 USD
102845-414
3,4-Dihydro-2H-1,4-benzoxazine-2-carbothioamide ≥97%
3,4-Dihydro-2H-1,4-benzoxazine-2-carbothioamide

Hazard: Irritant

Formula: C₉H₁₀N₂OS
MDL Number: MFCD09800603
CAS Number: 92659-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR