3,4-Dihydro-2H-1,4-benzoxazine-2-carbonitrile ≥97%
Supplier: Matrix Scientific
085576-1G
102845-410EA
226.15
USD
102845-410
3,4-Dihydro-2H-1,4-benzoxazine-2-carbonitrile ≥97%
3,4-Dihydro-2H-1,4-benzoxazine-2-carbonitrile
Hazard: Irritant
Formula:
C₉H₈N₂O |
MDL Number:
MFCD06200862 CAS Number: 86267-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...