2-Hydrazinoquinoxaline ≥97%

Supplier: Matrix Scientific

085574-10G 085574-1G
102845-406EA 1024.62 USD
102845-406 102845-404
2-Hydrazinoquinoxaline ≥97%
2-Hydrazinoquinoxaline

Hazard: Irritant

Formula: C₈H₈N₄
Melting Pt: 165…166 °C
MDL Number: MFCD00085165
CAS Number: 61645-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR