6-Nitroquinoxaline-2,3-dione ≥97%
Supplier: Matrix Scientific
085573-1G
102845-402EA
143.08
USD
102845-402
6-Nitroquinoxaline-2,3-dione ≥97%
6-Nitroquinoxaline-2,3-dione
Hazard: Irritant
Formula:
C₈H₃N₃O₄ MW: 205.13 g/mol |
MDL Number:
MFCD00673833 CAS Number: 2379-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...