Isoquinoline-1-carbonitrile ≥97%

Synonyms: 1-Isoquinolinecarbonitrile

085567-1G
102845-388EA 169.99 USD
102845-388
Isoquinoline-1-carbonitrile ≥97%
Isoquinoline-1-carbonitrile

Hazard: Irritant

Formula: C₁₀H₆N₂
MW: 154.17 g/mol
MDL Number: MFCD00134166
CAS Number: 1198-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR