2-Methyl-6-quinolinol ≥97%
Supplier: Matrix Scientific
Synonyms:
6-Hydroxy-2-methylquinoline
085563-1G
085563-10G
102845-376EA
1526.15
USD
102845-376
102845-374
2-Methyl-6-quinolinol ≥97%
2-Methyl-6-quinolinol
Hazard: Irritant
Formula:
C₁₀H₉NO MW: 159.19 g/mol |
MDL Number:
MFCD00047600 CAS Number: 613-21-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...