Decahydroquinoline ≥97%
Supplier: Matrix Scientific
085559-10G
085559-1G
102845-366EA
178.46
USD
102845-366
102845-364
Decahydroquinoline ≥97%
Decahydroquinoline
Hazard: Irritant
Formula:
C₉H₁₇N MW: 139.242 g/mol |
MDL Number:
MFCD00006695 CAS Number: 2051-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...