4-Nitro-1,3-dihydro-2H-benzimidazol-2-one ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Nitro-1H-benzo[d]imidazol-2(3H)-one

085550-1G
102845-332EA 750.77 USD
102845-332
4-Nitro-1,3-dihydro-2H-benzimidazol-2-one ≥97%
4-Nitro-1,3-dihydro-2H-benzimidazol-2-one

Hazard: Irritant

Formula: C₇H₅N₃O₃
MW: 179.14 g/mol
MDL Number: MFCD09835484
CAS Number: 85330-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR