5-Chloro-1,3-dihydro-2H-benzimidazol-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-1H-benzo[d]imidazol-2(3H)-one

085544-1G
102845-318EA 184.62 USD
102845-318
5-Chloro-1,3-dihydro-2H-benzimidazol-2-one ≥95%
5-Chloro-1,3-dihydro-2H-benzimidazol-2-one

Hazard: Irritant

Formula: C₇H₅ClN₂O
MW: 168.58 g/mol
MDL Number: MFCD00493866
CAS Number: 2034-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR