Sodium-2-mercapto-1H-benzimidazole-5-sulfonate ≥97%

Supplier: Matrix Scientific

085539-1G
102845-306EA 467.69 USD
102845-306
Sodium-2-mercapto-1H-benzimidazole-5-sulfonate ≥97%
Sodium-2-mercapto-1H-benzimidazole-5-sulfonate

Hazard: Irritant

Formula: C₇H₅N₂NaO₃S₂
MW: 252.25 g/mol
MDL Number: MFCD00142935
CAS Number: 53918-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR