6-Nitro-1,3-benzoxazol-2(3H)-one ≥97%
Supplier: Matrix Scientific
Synonyms:
6-Nitrobenzoxazole-2(3H)-one
085532-10G
085532-1G
102845-290EA
646.15
USD
102845-290
102845-288
6-Nitro-1,3-benzoxazol-2(3H)-one ≥97%
6-Nitro-1,3-benzoxazol-2(3H)-one
Hazard: Irritant
Formula:
C₇H₄N₂O₄ MW: 180.12 g/mol |
MDL Number:
MFCD00463755 CAS Number: 4694-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...