5-Methylbenzoxazole ≥97%
Supplier: Matrix Scientific
Synonyms:
5-Methylbenzo[d]oxazole
085527-25G
085527-5G
102845-272EA
153.85
USD
102845-272
102845-274
5-Methylbenzoxazole ≥97%
5-Methylbenzoxazole
Hazard: Irritant
Formula:
C₈H₇NO MW: 133.15 g/mol |
MDL Number:
MFCD00216938 CAS Number: 10531-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...