6-Ethoxy-2-mercaptobenzothiazole ≥97%
Supplier: Matrix Scientific
Synonyms:
6-Ethoxybenzo[d]thiazole-2(3H)-thione
085524-1G
085524-10G
085524-50G
102845-262EA
101.54
USD
102845-262
102845-266
102845-264
6-Ethoxy-2-mercaptobenzothiazole ≥97%
6-Ethoxy-2-mercaptobenzothiazole
Hazard: Irritant
Formula:
C₉H₉NOS₂ MW: 211.31 g/mol |
MDL Number:
MFCD00467715 CAS Number: 120-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...