2-(1,3-Benzothiazol-2-yl)acetonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(Benzo[d]thiazol-2-yl)acetonitrile

085523-50G
102845-260EA 543.95 USD
102845-260
2-(1,3-Benzothiazol-2-yl)acetonitrile ≥97%
2-(1,3-Benzothiazol-2-yl)acetonitrile

Hazard: Irritant

Formula: C₉H₆N₂S
MW: 174.23 g/mol
MDL Number: MFCD00051633
CAS Number: 56278-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR