6-(Trifluoromethyl)indoline ≥97%
Supplier: Matrix Scientific
085520-1G
102845-250EA
553.85
USD
102845-250
6-(Trifluoromethyl)indoline ≥97%
6-(Trifluoromethyl)indoline
Hazard: Irritant
Formula:
C₉H₈F₃N |
MDL Number:
MFCD01075225 CAS Number: 181513-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...