4,6-Difluoroisatin ≥97%

Supplier: Matrix Scientific

Synonyms: 4,6-Difluoro-1H-indole-2,3-dione

085518-1G
102845-244EA 61.54 USD
102845-244
4,6-Difluoroisatin ≥97%
4,6-Difluoroisatin

Hazard: Irritant

Formula: C₈H₃F₂NO₂
MW: 183.1100 g/mol
MDL Number: MFCD03618552
CAS Number: 126674-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR