5-(Trifluoromethyl)isatin ≥97%

Supplier: Matrix Scientific

085516-1G
102845-240EA 300 USD
102845-240
5-(Trifluoromethyl)isatin ≥97%
5-(Trifluoromethyl)isatin

Hazard: Irritant

Formula: C₉H₄F₃NO₂
MDL Number: MFCD01569510
CAS Number: 345-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR