4-Piperazin-1-yl-1H-indole dihydrochloride ≥97%

Supplier: Matrix Scientific

085514-1G
102845-236EA 863.08 USD
102845-236
4-Piperazin-1-yl-1H-indole dihydrochloride ≥97%
4-Piperazin-1-yl-1H-indole dihydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇Cl₂N₃
MDL Number: MFCD00829148
CAS Number: 255714-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR