5-(5-Methoxy-1H-indol-2-yl)-1,3,4-oxadiazol-2(3H)-one ≥97%
Supplier: Matrix Scientific
085512-1G
102845-232EA
524.62
USD
102845-232
5-(5-Methoxy-1H-indol-2-yl)-1,3,4-oxadiazol-2(3H)-one ≥97%
5-(5-Methoxy-1H-indol-2-yl)-1,3,4-oxadiazol-2(3H)-one
Hazard: Irritant
Formula:
C₁₁H₉N₃O₃ |
MDL Number:
MFCD09991832 CAS Number: 200061-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...