5-Methylbenzo[b]thiophene ≥98%

Supplier: Matrix Scientific

085501-1G
102845-198EA 95.38 USD
102845-198
5-Methylbenzo[b]thiophene ≥98%
5-Methylbenzo[b]thiophene

Hazard: Irritant

Formula: C₉H₈S
MDL Number: MFCD00052509
CAS Number: 14315-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR