1,2,4-Oxadiazole-3-carbothioamide ≥95%

Supplier: Matrix Scientific

085496-1G
102845-188EA 426.15 USD
102845-188
1,2,4-Oxadiazole-3-carbothioamide ≥95%
1,2,4-Oxadiazole-3-carbothioamide

Hazard: Irritant

Formula: C₃H₃N₃OS
MDL Number: MFCD00457756
CAS Number: 39512-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR