4-Chloro-6-(4-methylpiperazin-1-yl)-5-nitropyrimidine ≥97%

Supplier: Matrix Scientific

085455-1G
102845-088EA 666.15 USD
102845-088
4-Chloro-6-(4-methylpiperazin-1-yl)-5-nitropyrimidine ≥97%
4-Chloro-6-(4-methylpiperazin-1-yl)-5-nitropyrimidine

Hazard: Irritant

Formula: C₉H₁₂ClN₅O₂
MDL Number: MFCD02945691
CAS Number: 154418-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR