4-Aminopyrimidine-5-carbonitrile ≥97%

Supplier: Matrix Scientific

085443-1G
102845-062EA 58.46 USD
102845-062
4-Aminopyrimidine-5-carbonitrile ≥97%
4-Aminopyrimidine-5-carbonitrile

Hazard: Irritant

Formula: C₅H₄N₄
MW: 120.11 g/mol
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR