Pyrimidine-2-carbohydrazide ≥97%

Supplier: Matrix Scientific

085437-1G
102845-050EA 504.62 USD
102845-050
Pyrimidine-2-carbohydrazide ≥97%
Pyrimidine-2-carbohydrazide

Hazard: Irritant

Formula: C₅H₆N₄O
MDL Number: MFCD00508242
CAS Number: 87362-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR