2,3-Dicyano-5-methylpyrazine ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Methylpyrazine-2,3-dicarbonitrile5-Methyl-2,3-pyrazinedicarbonitrile

085422-50G
102845-018EA 849.23 USD
102845-018
2,3-Dicyano-5-methylpyrazine ≥97%
2,3-Dicyano-5-methylpyrazine

Hazard: Irritant

Formula: C₇H₄N₄
MW: 144.1300 g/mol
MDL Number: MFCD00191635
CAS Number: 52197-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR